siRNA Details
| Gene Symbol: Fbxw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025730.2 | NP_001020901.1 | 4 | 495 | ||
| XM_006233623.2 | XP_006233685.1 | 3 | 854 | |||
| XM_006233624.2 | XP_006233686.1 | 5 | 557 | |||
| XM_006233625.2 | XP_006233687.1 | 5 | 799 | |||
| XM_006233626.2 | XP_006233688.1 | 4 | 451 | |||
| XM_006233627.2 | XP_006233689.1 | 4 | 695 | |||
| XM_006233628.2 | XP_006233690.1 | 3 | 211 | |||
| GenBank mRNA | BC099179.1 | - | 4 | 498 | 7383052 | |

Fbxw5
Genomic Map