siRNA Details
| Gene Symbol: RGD1560470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233652.2 | XP_006233714.1 | 2 | 2848 | ||
| XM_006233653.2 | XP_006233715.1 | 3 | 2763 | |||
| XM_008761615.1 | XP_008759837.1 | 2 | 4183 | |||
| GenBank mRNA | FQ211897.1 | - | 3 | 1493 | ||

RGD1560470
Genomic Map