siRNA Details
| Gene Symbol: Wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039034.1 | NP_001034123.1 | 13 | 1243 | ||
| XM_006233831.2 | XP_006233893.1 | 14 | 1383 | |||
| GenBank mRNA | BC100156.1 | - | 13 | 1243 | 7462311 | |
| FQ222569.1 | - | 3 | 163 | |||

Wdr5
Genomic Map