siRNA Details
| Gene Symbol: Wdr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039034.1 | NP_001034123.1 | 4, 5 | 639 | ||
| XM_006233831.2 | XP_006233893.1 | 5, 6 | 779 | |||
| XM_008761692.1 | XP_008759914.1 | 780 | ||||
| GenBank mRNA | BC100156.1 | - | 4, 5 | 639 | 7462311 | |

Wdr5
Genomic Map