siRNA Details
Gene Symbol: Setx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233886.2 | XP_006233948.1 | 10 | 4345 | ||
XM_006233887.2 | XP_006233949.1 | 11 | 4348 | |||
XM_006233888.2 | XP_006233950.1 | 10 | 4344 | |||
XM_008761681.1 | XP_008759903.1 | 10 | 4459 | |||
XM_008761682.1 | XP_008759904.1 | 7 | 3713 |
