siRNA Details
| Gene Symbol: Setx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233886.2 | XP_006233948.1 | 10 | 3509 | ||
| XM_006233887.2 | XP_006233949.1 | 11 | 3512 | |||
| XM_006233888.2 | XP_006233950.1 | 10 | 3508 | |||
| XM_008761681.1 | XP_008759903.1 | 10 | 3623 | |||
| XM_008761682.1 | XP_008759904.1 | 7 | 2877 | |||

Setx
Genomic Map