siRNA Details
| Gene Symbol: Cdk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007743.1 | NP_001007744.1 | 7 | 1136 | ||
| GenBank mRNA | BC082037.1 | - | 7 | 1136 | 7130973 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk9| Gene Symbol: Cdk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007743.1 | NP_001007744.1 | 7 | 1136 | ||
| GenBank mRNA | BC082037.1 | - | 7 | 1136 | 7130973 | |