siRNA Details
| Gene Symbol: Wdsub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014179.1 | NP_001014201.1 | 5 | 958 | ||
| XM_006234208.2 | XP_006234270.1 | 5 | 1218 | |||
| XM_006234209.2 | XP_006234271.1 | 5 | 1217 | |||
| XM_006234211.2 | XP_006234273.1 | 5 | 939 | |||
| GenBank mRNA | BC089856.1 | - | 5 | 958 | 7381399 | |

Wdsub1
Genomic Map