siRNA Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077429.1 | NP_001070897.1 | 3 | 161 | ||
| XM_006234377.2 | XP_006234439.1 | 3 | 243 | |||
| XM_006234378.2 | XP_006234440.1 | 4 | 264 | |||
| XM_006234379.2 | XP_006234441.1 | 3 | 291 | |||
| XM_006234380.2 | XP_006234442.1 | 3 | 363 | |||
| XM_006234381.2 | XP_006234443.1 | 3 | 456 | |||
| XM_006234382.2 | XP_006234444.1 | 4 | 384 | |||
| XM_006234385.2 | XP_006234447.1 | 3 | 245 | |||
| XM_008761909.1 | XP_008760131.1 | 3 | 1340 | |||
| XM_008761910.1 | XP_008760132.1 | 3 | 245 | |||
| GenBank mRNA | BC126100.1 | - | 3 | 161 | 7132654 | |
| FQ218047.1 | - | 2 | 40 | |||

Lnp
Genomic Map