siRNA Details
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271212.1 | NP_001258141.1 | 12 | 1286 | ||
GenBank mRNA | CB774831.1 | - | 3 | 305 | ||
FQ217242.1 | - | 1288 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271212.1 | NP_001258141.1 | 12 | 1286 | ||
GenBank mRNA | CB774831.1 | - | 3 | 305 | ||
FQ217242.1 | - | 1288 |