siRNA Details
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271212.1 | NP_001258141.1 | 2 | 338 | ||
GenBank mRNA | FQ217242.1 | - | 340 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271212.1 | NP_001258141.1 | 2 | 338 | ||
GenBank mRNA | FQ217242.1 | - | 340 |