siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127379.1 | NP_001120851.1 | 12 | 1372 | ||
| XM_006234586.1 | XP_006234648.1 | 12 | 1465 | |||
| GenBank mRNA | BC162003.1 | - | 12 | 1372 | 9089120 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Api5| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127379.1 | NP_001120851.1 | 12 | 1372 | ||
| XM_006234586.1 | XP_006234648.1 | 12 | 1465 | |||
| GenBank mRNA | BC162003.1 | - | 12 | 1372 | 9089120 | |