siRNA Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013203.1 | NP_001013221.1 | 3 | 872 | ||
| GenBank mRNA | AY551091.1 | - | 3 | 872 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim44| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013203.1 | NP_001013221.1 | 3 | 872 | ||
| GenBank mRNA | AY551091.1 | - | 3 | 872 | ||