siRNA Details
| Gene Symbol: Caprin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012185.2 | NP_001012185.2 | 11 | 1235 | ||
| XM_006234652.2 | XP_006234714.1 | 12 | 1290 | |||
| GenBank mRNA | AB373991.1 | - | 10 | 1150 | ||
| AY155572.1 | - | 10 | 1091 | |||
| BC087123.1 | - | 10 | 1105 | |||

Caprin1
Genomic Map