siRNA Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013204.1 | NP_001013222.1 | 1 | 38 | ||
| XM_006234859.1 | XP_006234921.1 | 1 | 133 | |||
| GenBank mRNA | BC091256.1 | - | 1 | 38 | 7375652 | |
| FQ233751.1 | - | 1 | 74 | |||

Ccndbp1
Genomic Map