siRNA Details
| Gene Symbol: Sec23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277235.1 | NP_001264164.1 | 16 | 1838 | ||
| XM_006235126.2 | XP_006235188.1 | 17 | 2031 | |||
| XM_006235127.2 | XP_006235189.1 | 17 | 2020 | |||
| GenBank mRNA | FQ211753.1 | - | 16 | 1838 | ||
| FQ219827.1 | - | 2 | 71 | |||

Sec23b
Genomic Map