siRNA Details
| Gene Symbol: Naa20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108595.1 | NP_001102065.1 | 3 | 230 | ||
| GenBank mRNA | BC167020.1 | - | 3 | 190 | 9025632 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naa20| Gene Symbol: Naa20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108595.1 | NP_001102065.1 | 3 | 230 | ||
| GenBank mRNA | BC167020.1 | - | 3 | 190 | 9025632 | |