siRNA Details
| Gene Symbol: Naa20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108595.1 | NP_001102065.1 | 4 | 287 | ||
| XM_006235155.2 | XP_006235217.1 | 4 | 256 | |||
| GenBank mRNA | BC167020.1 | - | 4 | 247 | 9025632 | |
| FQ234868.1 | - | 4 | 212 | |||

Naa20
Genomic Map