siRNA Details
| Gene Symbol: Xrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108596.1 | NP_001102066.1 | 14 | 1524 | ||
| XM_006235156.2 | XP_006235218.1 | 15 | 1923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xrn2| Gene Symbol: Xrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108596.1 | NP_001102066.1 | 14 | 1524 | ||
| XM_006235156.2 | XP_006235218.1 | 15 | 1923 | |||