siRNA Details
Gene Symbol: Syndig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025020.1 | NP_001020191.1 | 4 | 751 | ||
GenBank mRNA | BC092131.1 | - | 4 | 751 | 7312153 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Syndig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025020.1 | NP_001020191.1 | 4 | 751 | ||
GenBank mRNA | BC092131.1 | - | 4 | 751 | 7312153 |