siRNA Details
| Gene Symbol: LOC100910990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749608.3 | XP_003749656.1 | 3 | 354 | ||
| XM_006235456.2 | XP_006235518.1 | 4 | 436 | |||
| XM_006235457.2 | XP_006235519.1 | 4 | 404 | |||
| XM_008762413.1 | XP_008760635.1 | 3 | 298 | |||
| XM_008762414.1 | XP_008760636.1 | 4 | 5512 | |||

LOC100910990
Genomic Map