siRNA Details
| Gene Symbol: Lpin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014184.1 | NP_001014206.1 | 1, 2 | 216 | ||
| XM_006235487.2 | XP_006235549.1 | 3, 4 | 1641 | |||
| XM_006235489.2 | XP_006235551.1 | 3, 4 | 992 | |||
| XM_006235490.2 | XP_006235552.1 | 4, 5 | 1786 | |||
| XM_006235491.2 | XP_006235553.1 | 3, 4 | 1252 | |||
| XM_006235493.2 | XP_006235555.1 | 2, 3 | 786 | |||
| XM_006235494.2 | XP_006235556.1 | 2, 3 | 1435 | |||
| GenBank mRNA | BC089878.1 | - | 1, 2 | 216 | 7326188 | |

Lpin3
Genomic Map