siRNA Details
| Gene Symbol: Ptprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108603.1 | NP_001102073.1 | 6 | 880 | ||
| XM_008762361.1 | XP_008760583.1 | 7 | 978 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprt| Gene Symbol: Ptprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108603.1 | NP_001102073.1 | 6 | 880 | ||
| XM_008762361.1 | XP_008760583.1 | 7 | 978 | |||