siRNA Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134955.1 | NP_001128427.1 | 21 | 2061 | ||
| XM_006235599.2 | XP_006235661.1 | 20 | 2037 | |||
| GenBank mRNA | BC089970.1 | - | 2 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elmo2| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134955.1 | NP_001128427.1 | 21 | 2061 | ||
| XM_006235599.2 | XP_006235661.1 | 20 | 2037 | |||
| GenBank mRNA | BC089970.1 | - | 2 | 530 | ||