siRNA Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134955.1 | NP_001128427.1 | 19 | 1868 | ||
| XM_006235599.2 | XP_006235661.1 | 18 | 1844 | |||
| Gene Symbol: LOC102555457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235631.2 | XP_006235693.1 | 18 | 1844 | ||

Elmo2
Genomic Map