siRNA Details
| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001517.1 | NP_001001517.1 | 5 | 592 | ||
| XM_008762515.1 | XP_008760737.1 | 5 | 1105 | |||
| GenBank mRNA | AY603473.1 | - | 5 | 592 | ||
| BC098633.1 | - | 5 | 583 | 7369070 | ||

Rnf114
Genomic Map