siRNA Details
| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214459.2 | NP_999624.1 | 6 | 942 | ||
| GenBank mRNA | AY500157.1 | - | 6 | 942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phactr3| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214459.2 | NP_999624.1 | 6 | 942 | ||
| GenBank mRNA | AY500157.1 | - | 6 | 942 | ||