siRNA Details
| Gene Symbol: Slc17a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108613.1 | NP_001102083.1 | 6 | 762 | ||
| XM_006235769.2 | XP_006235831.1 | 6 | 2701 | |||
| XM_006235771.2 | XP_006235833.1 | 6 | 2631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc17a9| Gene Symbol: Slc17a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108613.1 | NP_001102083.1 | 6 | 762 | ||
| XM_006235769.2 | XP_006235831.1 | 6 | 2701 | |||
| XM_006235771.2 | XP_006235833.1 | 6 | 2631 | |||