siRNA Details
| Gene Symbol: Sqrdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047913.2 | NP_001041378.2 | 2 | 383 | ||
| XM_006234887.2 | XP_006234949.1 | 3 | 455 | |||
| GenBank mRNA | AY325246.1 | - | 3 | 217 | ||
| BC107459.1 | - | 3 | 391 | |||
| BC158559.1 | - | 2 | 383 | 7456444 | ||

Sqrdl
Genomic Map