siRNA Details
| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012187.1 | NP_001012187.1 | 3, 4 | 332 | ||
| GenBank mRNA | BC083903.1 | - | 3, 4 | 332 | 7133606 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl7| Gene Symbol: Klhl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012187.1 | NP_001012187.1 | 3, 4 | 332 | ||
| GenBank mRNA | BC083903.1 | - | 3, 4 | 332 | 7133606 | |