siRNA Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014186.1 | NP_001014208.1 | 13, 14 | 1347 | ||
| XM_006235947.2 | XP_006236009.1 | 14, 15 | 1586 | |||
| GenBank mRNA | BC088447.1 | - | 13, 14 | 1347 | 7126516 | |

Orc5
Genomic Map