siRNA Details
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173554.1 | NP_001167025.1 | 5 | 1797 | ||
XM_006236090.2 | XP_006236152.1 | 4 | 1797 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mios | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173554.1 | NP_001167025.1 | 5 | 1797 | ||
XM_006236090.2 | XP_006236152.1 | 4 | 1797 |