siRNA Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105668.1 | NP_001099138.1 | 4 | 705 | ||
| XM_006236102.2 | XP_006236164.1 | 4 | 981 | |||
| XM_006236103.2 | XP_006236165.1 | 4 | 1010 | |||
| GenBank mRNA | CB325630.1 | - | 2 | 811 | ||
| CB785173.1 | - | 2 | 411 | |||
| CV106654.1 | - | 506 | ||||

Mdfic
Genomic Map