siRNA Details
| Gene Symbol: Flnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191862.1 | NP_001178791.1 | 2 | 567 | ||
| XM_006236214.2 | XP_006236276.1 | 2 | 723 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flnc| Gene Symbol: Flnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191862.1 | NP_001178791.1 | 2 | 567 | ||
| XM_006236214.2 | XP_006236276.1 | 2 | 723 | |||