siRNA Details
| Gene Symbol: Prss37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108625.1 | NP_001102095.1 | 4 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss37| Gene Symbol: Prss37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108625.1 | NP_001102095.1 | 4 | 527 | ||