siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108627.2 | NP_001102097.1 | 10 | 1365 | ||
| XM_006236409.1 | XP_006236471.1 | 9 | 1273 | |||
| XM_006236410.2 | XP_006236472.1 | 9 | 1275 | |||
| XM_006236411.1 | XP_006236473.1 | 9 | 1273 | |||
| GenBank mRNA | BC161932.1 | - | 10 | 1365 | 9021933 | |

Cul1
Genomic Map