siRNA Details
| Gene Symbol: Mpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134982.1 | NP_001128454.1 | 4 | 528 | ||
| XM_006236496.2 | XP_006236558.1 | 4 | 547 | |||
| XM_006236497.2 | XP_006236559.1 | 4 | 613 | |||
| XM_008762895.1 | XP_008761117.1 | 4 | 1028 | |||
| XM_008762897.1 | XP_008761119.1 | 4 | 567 | |||
| XM_008762898.1 | XP_008761120.1 | 4 | 1027 | |||
| GenBank mRNA | BC169025.1 | - | 4 | 528 | 9027949 | |

Mpp6
Genomic Map