siRNA Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013210.1 | NP_001013228.1 | 3 | 538 | ||
| GenBank mRNA | BC083746.1 | - | 3 | 538 | 7192246 | |
| FQ216652.1 | - | 3 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp14| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013210.1 | NP_001013228.1 | 3 | 538 | ||
| GenBank mRNA | BC083746.1 | - | 3 | 538 | 7192246 | |
| FQ216652.1 | - | 3 | 585 | |||