siRNA Details
| Gene Symbol: Fkbp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013210.1 | NP_001013228.1 | 3 | 506 | ||
| GenBank mRNA | BC083746.1 | - | 3 | 506 | 7192246 | |
| FQ216652.1 | - | 3 | 553 | |||
| Gene Symbol: LOC100911857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752244.3 | XP_003752292.3 | 394 | |||

Fkbp14
Genomic Map