siRNA Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762963.1 | XP_008761185.1 | 11, 12 | 1579 | ||
| GenBank mRNA | BC085921.1 | - | 11, 12 | 1364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herc6| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762963.1 | XP_008761185.1 | 11, 12 | 1579 | ||
| GenBank mRNA | BC085921.1 | - | 11, 12 | 1364 | ||