siRNA Details
| Gene Symbol: Antxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044249.2 | NP_001037714.1 | 2 | 568 | ||
| GenBank mRNA | BC131853.1 | - | 2 | 568 | 7320366 | |
| DQ789143.1 | - | 2 | 217 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Antxr1| Gene Symbol: Antxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044249.2 | NP_001037714.1 | 2 | 568 | ||
| GenBank mRNA | BC131853.1 | - | 2 | 568 | 7320366 | |
| DQ789143.1 | - | 2 | 217 | |||