siRNA Details
Gene Symbol: Slc25a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108638.1 | NP_001102108.1 | 2 | 246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108638.1 | NP_001102108.1 | 2 | 246 |