siRNA Details
| Gene Symbol: Irak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025422.1 | NP_001020593.1 | 12 | 1660 | ||
| XM_008763194.1 | XP_008761416.1 | 12 | 1704 | |||
| GenBank mRNA | BC098060.1 | - | 12 | 1660 | 7458113 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irak2| Gene Symbol: Irak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025422.1 | NP_001020593.1 | 12 | 1660 | ||
| XM_008763194.1 | XP_008761416.1 | 12 | 1704 | |||
| GenBank mRNA | BC098060.1 | - | 12 | 1660 | 7458113 | |