siRNA Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108648.1 | NP_001102118.1 | 1 | 113 | ||
| XM_006237409.2 | XP_006237471.1 | 1 | 502 | |||
| XM_006237410.2 | XP_006237472.1 | 1 | 503 | |||
| GenBank mRNA | BC158714.1 | - | 1 | 64 | 9088606 | |

Itfg2
Genomic Map