siRNA Details
| Gene Symbol: RGD1306151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108652.1 | NP_001102122.1 | 9 | 1339 | ||
| XM_006237532.1 | XP_006237594.1 | 9 | 1431 | |||
| XM_008763398.1 | XP_008761620.1 | 9 | 2247 | |||
| XM_008763399.1 | XP_008761621.1 | 9 | 2216 | |||

RGD1306151
Genomic Map