siRNA Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 4, 5 | 713 | ||
| XM_008763402.1 | XP_008761624.1 | 4, 5 | 692 | |||
| GenBank mRNA | AY426740.1 | - | 4, 5 | 713 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfr1op2| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 4, 5 | 713 | ||
| XM_008763402.1 | XP_008761624.1 | 4, 5 | 692 | |||
| GenBank mRNA | AY426740.1 | - | 4, 5 | 713 | ||