siRNA Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 7 | 1007 | ||
| GenBank mRNA | AY426739.1 | - | 6 | 893 | ||
| AY426740.1 | - | 7 | 1007 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgfr1op2| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 7 | 1007 | ||
| GenBank mRNA | AY426739.1 | - | 6 | 893 | ||
| AY426740.1 | - | 7 | 1007 | |||