siRNA Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025423.1 | NP_001020594.1 | 6 | 548 | ||
| XM_008763507.1 | XP_008761729.1 | 6 | 929 | |||
| XM_008763508.1 | XP_008761730.1 | 7 | 596 | |||
| XM_008763509.1 | XP_008761731.1 | 4 | 350 | |||
| XM_008763510.1 | XP_008761732.1 | 4 | 727 | |||
| GenBank mRNA | BC085821.1 | - | 6 | 551 | ||
| BC097945.1 | - | 6 | 548 | 7323001 | ||

Adhfe1
Genomic Map