siRNA Details
| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108655.1 | NP_001102125.1 | 2 | 537 | ||
| XM_006237872.2 | XP_006237934.1 | 2 | 2045 | |||
| GenBank mRNA | BC161902.1 | - | 2 | 526 | 8367141 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhf2| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108655.1 | NP_001102125.1 | 2 | 537 | ||
| XM_006237872.2 | XP_006237934.1 | 2 | 2045 | |||
| GenBank mRNA | BC161902.1 | - | 2 | 526 | 8367141 | |