siRNA Details
Gene Symbol: Ccne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108656.1 | NP_001102126.1 | 2, 3 | 134 | ||
XM_006237873.1 | XP_006237935.1 | 2, 3 | 678 | |||
XM_006237875.1 | XP_006237937.1 | 2, 3 | 144 |
